Batteries 4 C Accus
Leverancier: DELTA T
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A100028 A090005 A100025 A090011 A100027 A085003
478-9917EA 165 EUR
478-9917 DLTTA090005 DLTTA100025 DLTTA090011 DLTTA100027 DLTTA085003
Batteries 4 C Accus
Batterijen
All delta T 4 C Accus maintain a temperature betwene 2...8 °C.
  • The 4C element can be pre-cooled in the refrigerator at 2...4 °C
  • Ideal for transport as a set with our insulating containers
  • Closure: Aluminum composite foil and protective cap made of plastic
  • PE natural
  • Blue fluid
With the help of 4C elements, it can ensure that the cold chain disconnection phenomenon can be avoided during the transportation. This is done by using deltaT special coolant, which releases latent heat between 4 °C and 6 °C. Therefore, the product can be maintained at an almost constant temperature during transportation without active cooling.