Isovaleryl chloride 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
169280250 169281000 169285000
ACRO169280250EA 25.4 EUR
ACRO169280250 ACRO169281000 ACRO169285000
Isovaleryl chloride 98%
Isovaleryl chloride
Formule: C₅H₉ClO
Kookpunt: 115 °C (1013 hPa)
Dichtheid: 0,98 g/cm³
Vlampunt: 27 °C
MDL Nummer: MFCD00000738
CAS nummer: 108-12-3
EINECS: 203-552-2
UN: 2920
ADR: 8,II
Merck Index: 13,05253
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4170 (20°C, 589 nm)
GC ≥97.5 %