Isobutyl chloroformate 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
176180250 176181000 176185000
ACRO176180250EA 29.1 EUR
ACRO176180250 ACRO176181000 ACRO176185000
Isobutyl chloroformate 98%
Isobutyl chloroformate
Formule: C₅H₉ClO₂
Kookpunt: 128,7 °C (1013 hPa)
Smeltpunt: –80 °C
Dichtheid: 1,0475 g/cm³
Vlampunt: 30 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000642
CAS nummer: 543-27-1
EINECS: 208-840-1
UN: 3489
ADR: 6.1,I
Merck Index: 13,05156
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4060 to 1.4080 (20°C, 589 nm)
GC ≤1.0 % diisobutyl carbonate
Color scale ≤100 APHA
Free acid (HCl) ≤0.5 %