Benzyltributylammonium chloride 98+%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
186151000 186155000
ACRO186151000EA 27.5 EUR
ACRO186151000 ACRO186155000
Benzyltributylammonium chloride 98+%
Benzyltributylammonium chloride
Formule: C₁₉H₃₄ClN
Smeltpunt: 155…163 °C
MDL Nummer: MFCD00011849
CAS nummer: 23616-79-7
EINECS: 245-787-3
Order Now
Specification Test Results
Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Titration Argentometric 98.0 to 102.0 %
Water ≤1 %
HPLC ≥96.0 %