Tris(triphenylphosphine)rhodium(I) chloride 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Wilkinson's catalyst, Chlorotris(triphenylphosphine)rhodium(I), Tris(triphenylphosphine)chlororhodium
190420010 190420050
ACRO190420010EA 291 EUR
ACRO190420010 ACRO190420050
Tris(triphenylphosphine)rhodium(I) chloride 98%
Tris(triphenylphosphine)rhodium(I) chloride
Formule: C₅₄H₄₅ClP₃Rh
MW: 925.23 g/mol
Smeltpunt: 245…250 °C
MDL Nummer: MFCD00010016
CAS nummer: 14694-95-2
EINECS: 238-744-5
Merck Index: 12,02226
Order Now
Specification Test Results
Appearance Red-brown to brown Crystalline powder
Infrared spectrum Conforms
Gravimetry 10.8 to 11.3 % (Rh)