Allyltrimethylsilane 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

196990100 196990500 196992500
ACRO196990100EA 63.2 EUR
ACRO196990100 ACRO196990500 ACRO196992500
Allyltrimethylsilane 99%
Allyltrimethylsilane
Formule: H₂CCHCH₂Si(CH₃)₃
Kookpunt: 85 °C (760 mmHg)
Dichtheid: 0,719 g/cm³
Vlampunt: 7 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008635
CAS nummer: 762-72-1
EINECS: 212-104-5
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4060 to 1.4080 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR