n-Butyllithium 2.5 M in hexanes, AcroSeal®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
213351000 213358000 213350500
213351000.EA 63.4 EUR
213351000. 213358000. 213350500.
n-Butyllithium 2.5 M in hexanes, AcroSeal®
n-Butyllithium
Formule: C₄H₉Li
MW: 64.06 g/mol
Kookpunt: 60…80 °C (760 mmHg)
Smeltpunt: –95 °C
Dichtheid: 0.69 g/cm³
Vlampunt: –21 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009414
CAS nummer: 109-72-8
EINECS: 203-698-7
UN: 3394
ADR: 4.2
Order Now
Specification Test Results
Appearance Clear to hazy colorless to yellow Solution (May develop some turbidity or precipitate)
Titration Active base 2.37 to 2.59 M
Free base ≤1.2 %