1-Naphthalenesulphonyl chloride
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AC21664.50
AC21664.10
MAYBAC21664.50EA
0
EUR
MAYBAC21664.50
MAYBAC21664.10
1-Naphthalenesulphonyl chloride
1-Naphthalenesulphonyl chloride
Formule:
C₁₀H₇ClO₂S Kookpunt: 194 °C (1013 hPa) Smeltpunt: 67 °C |
MDL Nummer:
MFCD00003984 CAS nummer: 85-46-1 EINECS: 201-609-6 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...