2-Amino-2-ethyl-1,3-propanediol 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
227115000 227110025
ACRO227115000EA 93.4 EUR
ACRO227115000 ACRO227110025
2-Amino-2-ethyl-1,3-propanediol 97%
2-Amino-2-ethyl-1,3-propanediol
Formule: C₅H₁₃NO₂
Kookpunt: 152…153 °C (10 mmHg)
Smeltpunt: 35…38 °C
Dichtheid: 1,099 g/cm³
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004680
CAS nummer: 115-70-8
EINECS: 204-101-2
Merck Index: 12,00459
Order Now
Specification Test Results
Appearance Clear colorless to yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4850 to 1.4900 (20°C, 589 nm)
Titration with HClO4 ≥96.0 %