Occupational Shoes, Medical Clogs, OB SRC 87081 ACTIVE
Leverancier: Abeba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
87081/36 87081/37 87081/38 87081/39 87081/40 87081/41 87081/42 87081/43
ABEB87081/36EA 47.3 EUR
ABEB87081/36 ABEB87081/37 ABEB87081/38 ABEB87081/39 ABEB87081/40 ABEB87081/41 ABEB87081/42 ABEB87081/43
Occupational Shoes, Medical Clogs, OB SRC 87081 ACTIVE
Schoenen en laarzen
Abeba® 87081 ACTIVE are comfortable, lightweight clogs made of washable grain leather that are shoes specially designed to reduce leg fatigue and protect against injuries during long hours of standing at work. Ideal for hospitals, nursing homes and the food industry.
  • Increased breathability
  • Increased durability of the upper grain leather
  • Washable at 30 °C
  • Slip-resistant outer sole with self-cleaning technology
  • Resistance to brief contact with hot substances up to 180 °C
  • Lightweight; Antistatic
Energy absorption in the heel area and sole is resistantMore Product Information