Isobutyric anhydride 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
257780020 257785000
ACRO257780020EA 240 EUR
ACRO257780020 ACRO257785000
Isobutyric anhydride 97%
Isobutyric anhydride
Formule: C₈H₁₄O₃
Kookpunt: 183…187 °C (1013 hPa)
Smeltpunt: –53 °C
Dichtheid: 0,954 g/cm³
MDL Nummer: MFCD00008913
CAS nummer: 97-72-3
EINECS: 202-603-6
UN: 2922
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4050 to 1.4070 (20°C, 589 nm)
GC ≤1.0 % (Isobutyric acid)
Propionyl content ≤1.0 % (as propionic acid)