Benzoyl isocyanate 90% tech.
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
314280050 314280010
ACRO314280050EA 0 EUR
ACRO314280050 ACRO314280010
Benzoyl isocyanate 90% tech.
Benzoyl isocyanate
Formule: C₈H₅NO₂
Kookpunt: 94…96 °C (21 mmHg)
Smeltpunt: 28…29 °C
Dichtheid: 1,17 g/cm³
Vlampunt: 72 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00002032
CAS nummer: 4461-33-0
EINECS: 224-717-5
UN: 2206
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear to hazy colorless to yellow Liquid or solid
Infrared spectrum Conforms
GC ≥88 %