6-Aminonicotinonitrile 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
357620050 357620250
ACRO357620050EA 67.4 EUR
ACRO357620050 ACRO357620250
6-Aminonicotinonitrile 98%
6-Aminonicotinonitrile
Formule: C₆H₅N₃
Kookpunt: 240…250 °C (15 mmHg)
Smeltpunt: 159…165 °C
MDL Nummer: MFCD00128902
CAS nummer: 4214-73-7
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Brown Crystalline powder
Infrared spectrum Conforms
Melting point 159 to 165 °C
GC ≥97.5 %