Tri-tert-butylphosphine 95%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Tri-t-butylphosphine, Tris(1,1-dimethylethyl)phosphine
360890010 360890050
ACRO360890010EA 89.8 EUR
ACRO360890010 ACRO360890050
Tri-tert-butylphosphine 95%
Tri-tert-butylphosphine
Formule: C₁₂H₂₇P
MW: 202.32 g/mol
Kookpunt: 102…103 °C (13 mmHg)
Smeltpunt: 30 °C
Vlampunt: –17 °C
MDL Nummer: MFCD00015006
CAS nummer: 13716-12-6
EINECS: 237-266-4
UN: 2846
ADR: 4.2 I
Order Now
Specification Test Results
Appearance Colorless Low melting solid
Infrared spectrum Conforms
NMR ≥94 %