n-Butyllithium 2.7 M in toluene, AcroSeal®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
378931000 378938000
378931000.EA 44.4 EUR
378931000. 378938000.
n-Butyllithium 2.7 M in toluene, AcroSeal®
n-Butyllithium
Formule: C₄H₉Li
MW: 64.06 g/mol
Smeltpunt: -76
Dichtheid: 0.86 g/cm³
Vlampunt: 4 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009414
CAS nummer: 109-72-8
EINECS: 203-698-7
UN: 3394
ADR: 4.2
Order Now
Specification Test Results
Appearance Yellow to orange Solution (May develop some turbidity or precipitate)
Titration Active base 2.62 to 2.75 M
Free base ≤1.0 %