N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylsilyl chloride
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
389320250 389321000 389325000
ACRO389320250EA 228 EUR
ACRO389320250 ACRO389321000 ACRO389325000
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylsilyl chloride
N,O-Bis(trimethylsilyl)trifluoroacetamide
Formule: C₈H₁₈F₃NOSi₂
Kookpunt: 145…147 °C (760 mmHg)
Smeltpunt: -41 °C
Dichtheid: 0,974 g/cm³
Vlampunt: 34 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008269
CAS nummer: 25561-30-2
EINECS: 247-103-9
UN: 2920
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
GC ≤1.5 % TMCS