Calcium formate 98%, pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
403830010 403832500 403830025
ACRO403830010EA 151 EUR
ACRO403830010 ACRO403832500 ACRO403830025
Calcium formate 98%, pure
Calcium formate
Formule: C₂H₂CaO₄
Smeltpunt: >300 °C
Dichtheid: 2,009 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00036108
CAS nummer: 544-17-2
EINECS: 208-863-7
Merck Index: 12,01711
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %
Stabilizer 2 % CaCO3
Specific optical rotation +200° to +220° (20°C, 589 nm) (c=3, CHCl3)