4-Nitrophthalic acid 99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
416120250
416121000
ACRO416120250EA
186
EUR
ACRO416120250
ACRO416121000
4-Nitrophthalic acid 99%
4-Nitrophthalic acid
Formule:
C₈H₅NO₆ Smeltpunt: 165 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00007252 CAS nummer: 610-27-5 EINECS: 210-215-3 |
Specification Test Results
Appearance | Yellow to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 163 to 167 °C |
HPLC | ≥98.5 % |
Titration with TBAH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...