Kaliumpolysulfiden (≥11,8% S)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Potassium polysulphide, Liver of sulphur, Sulphurated potash
41550.22 41550.36
41550.22EA 63.2 EUR
41550.22 41550.36
Kaliumpolysulfiden (≥11,8% S)
Kaliumpolysulfiden
CAS No.: 39365-88-3 Molecular Formula: K2Sn Sensitivity: Air Sensitive and Hygroscopic Solubility: Soluble in water UN No.: 3180 MDL No.: MFCD00064604
Decorative Colour effects on brass, bronze, nickel
Formule: K₂(S)n
MW: 110.26 g/mol
Smeltpunt: 471
Dichtheid: 1.65 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064604
CAS nummer: 37199-66-9
EINECS: 253-390-1
UN: 3180
ADR: 4.1 II