Pentaerythritol ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A10520.0B
A10520.0I
A10520.30
A10520.0BEA
38
EUR
A10520.0B
A10520.0I
A10520.30
Pentaerythritol ≥98%
Pentaerythritol
Purity: 98+%
CAS No.: 115-77-5
Molecular Formula: C5H12O4
Formula Weight: 136.15
Flash Point: 240°(464°F)
Melting Point: 260-264°
Boiling Point: 276°/30mm
Beilstein Reference: 1679274
Merck Reference: 14,7111
MDL No.: MFCD00004692
Formule:
C₅H₁₂O₄ Kookpunt: 276 °C (30 mmHg) Smeltpunt: 260…264 °C Dichtheid: 1,396 Vlampunt: 240 °C (464 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004692 CAS nummer: 115-77-5 EINECS: 204-104-9 Merck Index: 14,7111 |