Cyanoacetic acid ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
A11045.0B
A11045.0I
A11045.30
A11045.0BEA
120
EUR
A11045.0B
A11045.0I
A11045.30
Cyanoacetic acid ≥97%
Cyanoacetic acid
Purity: 99%
CAS No.: 372-09-8
Molecular Formula: NCCH2CO2H
Formula Weight: 85.06
Flash Point: 107°(224°F)
Odour: Odourless
pH: 1.5
Melting Point: 64-69°
Boiling Point: 108°/15mm
Beilstein Reference: 506325
Merck Reference: 14,2689
UN No.: 3261
MDL No.: MFCD00002677
Formule:
C₃H₃NO₂ MW: 85.06 g/mol Kookpunt: 108 °C (15 mmHg) Smeltpunt: 64…69 °C Dichtheid: Vlampunt: 107 °C (224 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002677 CAS nummer: 372-09-8 EINECS: 206-743-9 UN: 3261 ADR: 8 II Merck Index: 13,02718 |