Butyltrichlorosilane ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
A11256.06
A11256.14
A11256.22
A11256.06EA
24.8
EUR
A11256.06
A11256.14
A11256.22
Butyltrichlorosilane ≥97%
Butyltrichlorosilane
Purity: 97+%
CAS No.: 7521-80-4
Molecular Formula: C4H9Cl3Si
Formula Weight: 191.56
Flash Point: 45°(113°F)
Refractive Index: 1.4370
Odour: Acrid
Boiling Point: 142-143°
Beilstein Reference: 1736032
UN No.: 1747
MDL No.: MFCD00013605
Formule:
C₄H₉Cl₃Si Kookpunt: 142…143 °C Dichtheid: 1,160 Vlampunt: 45 °C (113 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00013605 CAS nummer: 7521-80-4 EINECS: 231-381-3 UN: 1747 ADR: 8,II |