4-Fluoro-o-phenylenediamine 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
A13977.06 A13977.14 A13977.22
A13977.06EA 69.9 EUR
A13977.06 A13977.14 A13977.22
4-Fluoro-o-phenylenediamine 97%
4-Fluoro-o-phenylenediamine
Purity: 97% CAS No.: 367-31-7 Molecular Formula: C6H7FN2 Formula Weight: 126.13 Melting Point: 91-95° Beilstein Reference: 2081075 UN No.: 2811 MDL No.: MFCD00042228
Formule: C₆H₇FN₂
Kookpunt: 250 °C (1013 hPa)
Smeltpunt: 91…95 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00042228
CAS nummer: 367-31-7
EINECS: 206-691-7
UN: 2811
ADR: 6.1,III
Order Now
Specificaties
Environmentally Preferable