trans-Cinnamaldehyde ≥98%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Purity: 98+%
CAS No.: 14371-10-9
Molecular Formula: C9H8O
Formula Weight: 132.16
Flash Point: 131°(268°F)
Physical Form: Liquid
Density: 1.050
Refractive Index: 1.6220
Sensitivity: Air Sensitive
Appearance: Yellow
Odour: Characteristic
Melting Point: -8°
Boiling Point: 250-252°
Beilstein Reference: 1071571
Merck Reference: 14,2297
MDL No.: MFCD00007000
Formule:
C₉H₈O Kookpunt: 250…252 °C Smeltpunt: –8 °C Dichtheid: 1,050 Vlampunt: 131 °C (268 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00007000 CAS nummer: 14371-10-9 Merck Index: 14,2297 |