Ethylchlooracetaat ≥99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
A15554.0E
A15554.22
A15554.36
A15554.0EEA
113
EUR
A15554.0E
A15554.22
A15554.36
Ethylchlooracetaat ≥99%
Ethylchlooracetaat
Purity: 99%
CAS No.: 105-39-5
Molecular Formula: C4H7ClO2
Formula Weight: 122.55
Flash Point: 54°(149°F)
Refractive Index: 1.4210
Odour: Pungent
Melting Point: -26°
Boiling Point: 142-144°
Beilstein Reference: 506455
Merck Reference: 14,3783
UN No.: 1181
MDL No.: MFCD00000932
Formule:
C₄H₇ClO₂ Kookpunt: 142…144 °C Smeltpunt: –26 °C Dichtheid: 1,159 Vlampunt: 54 °C (149 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000932 CAS nummer: 105-39-5 EINECS: 203-294-0 UN: 1181 ADR: 6.1,II Merck Index: 14,3783 |