(±)-Tetrahydro-2-furfurylmethanol 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: (±)-Tetrahydro-2-furfuryl alcohol, (Tetrahydrofuran-2-yl)methanol, (±)-Tetrahydro-2-furyl methanol, (±)-THFA
A17078.0F A17078.AE A17078.AP
A17078.0FEA 78.6 EUR
A17078.0F A17078.AE A17078.AP
(±)-Tetrahydro-2-furfurylmethanol 98%
(±)-Tetrahydro-2-furfurylmethanol
Purity: 98% CAS No.: 97-99-4 Molecular Formula: C5H10O2 Formula Weight: 102.13 Flash Point: 75°(167°F) Refractive Index: 1.4520 Melting Point: -80° Boiling Point: 178° Beilstein Reference: 102723 Merck Reference: 14,9213 MDL No.: MFCD00005372
Formule: C₅H₁₀O₂
MW: 102.13 g/mol
Kookpunt: 178 °C
Smeltpunt: < –80 °C
Dichtheid: 1.053
Vlampunt: 75 °C (167 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005372
CAS nummer: 97-99-4
EINECS: 202-625-6
Merck Index: 12,09353
Order Now
Specificaties
Environmentally Preferable