Dual Stage Gas Regulators
Leverancier: DISTEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5000-2009 5000-2010 5000-2011 5000-2012
DISI5000-2009EA 0 EUR
DISI5000-2009 DISI5000-2010 DISI5000-2011 DISI5000-2012
Dual Stage Gas Regulators
Gasregelaars
Dual stage gas regulator with outlet connection - ¹/₄" tube fitting.
  • Delivery range - 0 to 3,44 bar