1H,1H,2H,2H-Perfluoro-1-iodooctane ≥97% stabilised
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodooctane, 1H,1H,2H,2H-Perfluorooctyl iodide, 1-Iodo-3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctane
Stabilisatie:
Stabilised with copper
B20984.06
B20984.14
B20984.06EA
41.8
EUR
B20984.06
B20984.14
1H,1H,2H,2H-Perfluoro-1-iodooctane ≥97% stabilised
1H,1H,2H,2H-Perfluoro-1-iodooctane
Stabilized with copper
Purity: 97%
CAS No.: 2043-57-4
Molecular Formula: CF3(CF2)5CH2CH2I
Formula Weight: 474.00
Flash Point: 150°(302°F)
Refractive Index: 1.3600
Melting Point: ca 20°
Boiling Point: 178-180°
Beilstein Reference: 1887105
MDL No.: MFCD00039410
Formule:
C₈H₄F₁₃I Kookpunt: 178…180 °C Smeltpunt: ∼ 20 °C Dichtheid: 1,940 Vlampunt: 150 °C (302 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00039410 CAS nummer: 2043-57-4 EINECS: 218-056-1 |