Butyl vinyl ether ≥98% stabilised
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Stabilisatie:
Stabilised with potassium hydroxide 100 ppm
B23673.AE
B23673.AP
B23673.AEEA
26
EUR
B23673.AE
B23673.AP
Butyl vinyl ether ≥98% stabilised
Butyl vinyl ether
Stabilized with 0.01% KOH
Purity: 98%
CAS No.: 111-34-2
Molecular Formula: C6H12O
Formula Weight: 100.16
Storage Temperature: 2°C to 8°C
Flash Point: -12°(10°F)
Refractive Index: 1.4010
Melting Point: -113°
Boiling Point: 92-94°
Beilstein Reference: 1560217
UN No.: 3271
MDL No.: MFCD00009454
Formule:
C₆H₁₂O Kookpunt: 92…94 °C Smeltpunt: –113 °C Dichtheid: 0,778 Vlampunt: –12 °C (10 °F) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00009454 CAS nummer: 111-34-2 EINECS: 203-860-7 UN: 3271 ADR: 3,II |