5-Hydroxyindole-2-carboxylic acid ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
5-Hydroxy-2-indolecarboxylic acid, 5-Hydroxy-1H-indole-2-carboxylic acid
L00786.03
L00786.06
L00786.03EA
63
EUR
L00786.03
L00786.06
5-Hydroxyindole-2-carboxylic acid ≥97%
5-Hydroxyindole-2-carboxylic acid
Purity: 97%
CAS No.: 21598-06-1
Molecular Formula: C9H7NO3
Formula Weight: 177.16
Storage Temperature: 2°C to 8°C
Melting Point: ca 245° dec.
Beilstein Reference: 153214
MDL No.: MFCD00005615
Formule:
C₉H₇NO₃ |
MDL Nummer:
MFCD00005615 CAS nummer: 21598-06-1 EINECS: 244-468-6 |