Tetrakis(triphenylphosphine)palladium(0) ≥99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Pd(PPh3)4
L00953.03 L00953.06
L00953.03EA 68.9 EUR
L00953.03 L00953.06
Tetrakis(triphenylphosphine)palladium(0) ≥99%
Tetrakis(triphenylphosphine)palladium(0)
Formule: C₇₂H₆₀P₄Pd
MW: 1155.58 g/mol
Smeltpunt: 103…107 °C
Dichtheid: 1 g/cm³ (20 °C)
MDL Nummer: MFCD00010012
CAS nummer: 14221-01-3
EINECS: 238-086-9