(2-Methoxyethoxy)methyl chloride ≥94%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
L01050.06
L01050.14
L01050.06EA
41.8
EUR
L01050.06
L01050.14
(2-Methoxyethoxy)methyl chloride ≥94%
(2-Methoxyethoxy)methyl chloride
Purity: 94%
CAS No.: 3970-21-6
Molecular Formula: C4H9ClO2
Formula Weight: 124.57
Storage Temperature: -30°C to -10°C
Flash Point: 54°(129°F)
Refractive Index: 1.4270
Boiling Point: 50-52°/13mm
Beilstein Reference: 1900576
UN No.: 1992
MDL No.: MFCD00000888
Formule:
ClCH₂OCH₂CH₂OCH₃ Kookpunt: 50…52 °C (13 mmHg) Dichtheid: 1,094 Vlampunt: 54 °C (129 °F) |
MDL Nummer:
MFCD00000888 CAS nummer: 3970-21-6 EINECS: 223-589-8 UN: 1992 ADR: 3,III |