Tetrahydro-1,3-dimethyl-1H-pyrimidine-2-on ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Dimethylpropyleneurea, Dimethyl propylene urea, 1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone, 1,3-Dimethyltetrahydropyrimidin-2(1H)-one, DMPU, 1,3-Dimethyltetrahydro-2(1H)-pyrimidinone, N,N'-Dimethyl propylene urea
L09968.14
L09968.22
L09968.36
L09968.14EA
25.2
EUR
L09968.14
L09968.22
L09968.36
Tetrahydro-1,3-dimethyl-1H-pyrimidine-2-on ≥98%
Tetrahydro-1,3-dimethyl-1H-pyrimidine-2-on
Purity: 98%
CAS No.: 7226-23-5
Molecular Formula: C6H12N2O
Formula Weight: 128.18
Flash Point: 120°(248°F)
Refractive Index: 1.4890
Odour: Odourless
pH: 11
Melting Point: -23°
Boiling Point: 246-247°
Beilstein Reference: 110562
MDL No.: MFCD00006550
Formule:
C₆H₁₂N₂O MW: 128.17 g/mol Kookpunt: 246…247 °C Smeltpunt: –23 °C Dichtheid: 1.060 Vlampunt: 120 °C (248 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00006550 CAS nummer: 7226-23-5 EINECS: 230-625-6 |