4-Cyanophenyl isothiocyanate ≥98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 4-Cyanophenylisothiocyanate, 1-Cyano-4-isothiocyanatobenzene, 4-Isothiocyanatobenzonitrile
L10173.03 L10173.06
L10173.03EA 30.3 EUR
L10173.03 L10173.06
4-Cyanophenyl isothiocyanate ≥98%
4-Cyanophenyl isothiocyanate
Purity: 98% CAS No.: 2719-32-6 Molecular Formula: C8H4N2S Formula Weight: 160.20 Odour: Irritating Melting Point: 118-122° Beilstein Reference: 2802621 UN No.: 2923 MDL No.: MFCD00041085
Formule: C₈H₄N₂S
MW: 160.2 g/mol
Kookpunt: 294
Smeltpunt: 118…122 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00041085
CAS nummer: 2719-32-6
EINECS: 220-323-2
UN: 2923
ADR: 8 III