3-Chloro-4-fluorophenyl isocyanate ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
L10891.06
L10891.03
L10891.14
L10891.06EA
80.8
EUR
L10891.06
L10891.03
L10891.14
3-Chloro-4-fluorophenyl isocyanate ≥98%
3-Chloro-4-fluorophenyl isocyanate
Purity: 98%
CAS No.: 50529-33-4
Molecular Formula: C7H3ClFNO
Formula Weight: 171.56
Storage Temperature: 2°C to 8°C
Flash Point: 87°(188°F)
Refractive Index: 1.5410
Boiling Point: 44°/17mm
Beilstein Reference: 973324
UN No.: 2206
MDL No.: MFCD00037037
Formule:
C₇H₃ClFNO Kookpunt: 44 °C (17 mmHg) Dichtheid: 1,377 Vlampunt: 87 °C (188 °F) |
MDL Nummer:
MFCD00037037 CAS nummer: 50529-33-4 ADR: 6.1,II |