3-Chloro-4-methylphenyl isocyanate ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
L12064.06
L12064.14
L12064.03
L12064.06EA
42.9
EUR
L12064.06
L12064.14
L12064.03
3-Chloro-4-methylphenyl isocyanate ≥98%
3-Chloro-4-methylphenyl isocyanate
Purity: 98%
CAS No.: 28479-22-3
Molecular Formula: C8H6ClNO
Formula Weight: 167.60
Storage Temperature: 2°C to 8°C
Flash Point: 109°(228°F)
Refractive Index: 1.5560
Melting Point: 20-24°
Boiling Point: 225°
Beilstein Reference: 2690771
UN No.: 2236
MDL No.: MFCD00013858
Formule:
C₈H₆ClNO Kookpunt: 225 °C Smeltpunt: 20…24 °C Dichtheid: 1,224 Vlampunt: 109 °C (228 °F) |
MDL Nummer:
MFCD00013858 CAS nummer: 28479-22-3 EINECS: 249-050-7 UN: 2236 ADR: 6.1,II |