Benzenesulphonic acid ∼75% (w/w) in aqueous solution, Technical Grade
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
L19077.AK L19077.AU L19077.0J
L19077.AKEA 22.1 EUR
L19077.AK L19077.AU L19077.0J
Benzenesulphonic acid ∼75% (w/w) in aqueous solution, Technical Grade
Benzenesulphonic acid
CAS No.: 98-11-3 Molecular Formula: C6H6O3S Formula Weight: 158.18 Flash Point: 110°(230°F) Beilstein Reference: 742513 Merck Reference: 14,1070 MDL No.: MFCD00011689
Formule: C₆H₆O₃S
MW: 158.18 g/mol
Kookpunt: 319
Smeltpunt: 50
Dichtheid: 1.30
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011689
CAS nummer: 98-11-3
EINECS: 202-638-7
UN: 2586
ADR: 8 III
Merck Index: 14,1070