7-Nitro-1H-indazole
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
81340-100 81340-50 81340-250 81340-500
CAYM81340-100EA 92.5 EUR
CAYM81340-100 CAYM81340-50 CAYM81340-250 CAYM81340-500
7-Nitro-1H-indazole
7-Nitro-1H-indazole
Formule: C₇H₅N₃O₂
Smeltpunt: 190 °C
Dichtheid: 1,56 g/cm³ (293 °C)
MDL Nummer: MFCD00022789
CAS nummer: 2942-42-9
EINECS: 220-934-4