5'-Fluoro-2'-hydroxyacetophenone

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 1-(5-Fluoro-2-hydroxyphenyl)ethanone

BTB04130.1 BTB04130EA BTB04130EE
MAYBBTB04130.1EA 0 EUR
MAYBBTB04130.1 MAYBBTB04130EA MAYBBTB04130EE
5'-Fluoro-2'-hydroxyacetophenone
5'-Fluoro-2'-hydroxyacetophenone
Formule: C₈H₇FO₂
MW: 154.14 g/mol
Kookpunt: 182
Smeltpunt: 56
MDL Nummer: MFCD00011668
CAS nummer: 394-32-1
EINECS: 206-893-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR