1-(4-Methoxyphenyl)-2-thiourea
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
BTB06797.1 BTB06797.10
MAYBBTB06797.1EA 0 EUR
MAYBBTB06797.1 MAYBBTB06797.10
1-(4-Methoxyphenyl)-2-thiourea
1-(4-Methoxyphenyl)-2-thiourea
Formule: (C₆H₄(OCH₃))NC(S)NH₂
MDL Nummer: MFCD00004936
CAS nummer: 2293-07-4
EINECS: 218-931-8
UN: 2811
ADR: 6.1,III