5-Chloro-o-anisidine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 5-Chloro-2-methoxyaniline, 5-Chloro-2-anisidine, 2-Amino-4-chloroanisole

BTB09115.1 BTB09115.10 BTB09115.50
MAYBBTB09115.1EA 0 EUR
MAYBBTB09115.1 MAYBBTB09115.10 MAYBBTB09115.50
5-Chloro-o-anisidine
5-Chloro-o-anisidine
Formule: C₇H₈ClNO
MW: 157.6 g/mol
Kookpunt: 259
Smeltpunt: 84
MDL Nummer: MFCD00007777
CAS nummer: 95-03-4
EINECS: 202-385-2
UN: 2233
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR