5-Bromo-2-thenoic acid

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BTB12643.1 BTB12643.10
MAYBBTB12643.1EA 0 EUR
MAYBBTB12643.1 MAYBBTB12643.10
5-Bromo-2-thenoic acid
5-Bromo-2-thenoic acid
Formule: C₅H₃BrO₂S
MDL Nummer: MFCD00079725
CAS nummer: 7311-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR