(Methyl)triphenylphosphonium iodide
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Methyltriphenylphosphonium iodide
CD03492.1
CD03492.10
CD03492.50
MAYBCD03492.1EA
0
EUR
MAYBCD03492.1
MAYBCD03492.10
MAYBCD03492.50
(Methyl)triphenylphosphonium iodide
(Methyl)triphenylphosphonium iodide
Formule:
C₁₉H₁₈IP Smeltpunt: 193…194 °C |
MDL Nummer:
MFCD00066175 CAS nummer: 2065-66-9 EINECS: 218-178-5 UN: 3464 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...