4-Cyanophenyl isocyanate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CD10847.1 CD10847.10 CD10847.50
MAYBCD10847.1EA 0 EUR
MAYBCD10847.1 MAYBCD10847.10 MAYBCD10847.50
4-Cyanophenyl isocyanate
4-Cyanophenyl isocyanate
Formule: C₈H₄N₂O
Kookpunt: 123 °C (12 mmHg)
Smeltpunt: 99 °C
CAS nummer: 40465-45-0
EINECS: 254-934-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR