3-Chloro-4-methylphenyl isocyanate
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CD11311.1 CD11311.10 CD11311.50
MAYBCD11311.1EA 0 EUR
MAYBCD11311.1 MAYBCD11311.10 MAYBCD11311.50
3-Chloro-4-methylphenyl isocyanate
3-Chloro-4-methylphenyl isocyanate
Formule: C₈H₆ClNO
Kookpunt: 225 °C (1013 hPa)
Dichtheid: 1,224 g/cm³ (20 °C)
MDL Nummer: MFCD00013858
CAS nummer: 28479-22-3
EINECS: 249-050-7
UN: 2236
ADR: 6.1,II