4-Isopropylbenzyl alcohol

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

SB01074.1 SB01074.10 SB01074.50
MAYBSB01074.1EA 0 EUR
MAYBSB01074.1 MAYBSB01074.10 MAYBSB01074.50
4-Isopropylbenzyl alcohol
4-Isopropylbenzyl alcohol
Formule: C₁₀H₁₄O
Kookpunt: 249 °C (1013 hPa)
Smeltpunt: 28 °C
Dichtheid: 0,978 g/cm³ (20 °C)
MDL Nummer: MFCD00004663
CAS nummer: 536-60-7
EINECS: 208-640-4
Merck Index: 13,02646

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR