4-Fluor-3-trifluormethylfenol

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SB01726.1 SB01726.10
MAYBSB01726.1EA 0 EUR
MAYBSB01726.1 MAYBSB01726.10
4-Fluor-3-trifluormethylfenol
4-Fluor-3-trifluormethylfenol
Formule: C₇H₄F₄O
Kookpunt: 84 °C (1013 hPa)
MDL Nummer: MFCD00043875
CAS nummer: 61721-07-1
EINECS: 432-560-0
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR