4-Butylphenyl isocyanate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SB01778.1 SB01778.10 SB01778.50
MAYBSB01778.1EA 0 EUR
MAYBSB01778.1 MAYBSB01778.10 MAYBSB01778.50
4-Butylphenyl isocyanate
4-Butylphenyl isocyanate
Formule: C₁₁H₁₃NO
Kookpunt: 121 °C (1013 hPa)
Dichtheid: 0,992 g/cm³ (20 °C)
MDL Nummer: MFCD00013882
CAS nummer: 69342-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR