tert-Butyl bromoacetate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SB01797.1 SB01797.10 SB01797.50
MAYBSB01797.1EA 0 EUR
MAYBSB01797.1 MAYBSB01797.10 MAYBSB01797.50
tert-Butyl bromoacetate
tert-Butyl bromoacetate
Formule: BrCH₂COOC(CH₃)₂
Kookpunt: 50 °C (1013 hPa)
Dichtheid: 1,338 g/cm³ (20 °C)
MDL Nummer: MFCD00000188
CAS nummer: 5292-43-3
EINECS: 226-133-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR