2-Methoxy-3-(trifluoromethyl)pyridine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SPB05753.1 SPB05753.10 SPB05753.50
MAYBSPB05753.1EA 0 EUR
MAYBSPB05753.1 MAYBSPB05753.10 MAYBSPB05753.50
2-Methoxy-3-(trifluoromethyl)pyridine
2-Methoxy-3-(trifluoromethyl)pyridine
Formule: C₇H₆F₃NO
MDL Nummer: MFCD00153202
CAS nummer: 121643-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR